Skip to Content
Merck
All Photos(1)

Key Documents

BP1182

Salmeterol impurity 1

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

1-Hydroxy-4-[2-hydroxy-5-(1-hydroxy-2-{[6-(4-phenylbutoxy)hexyl]amino}ethyl)benzyl]-2-naphthoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€200.00

€200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€200.00

About This Item

Empirical Formula (Hill Notation):
C36H43NO6
CAS Number:
Molecular Weight:
585.73
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€200.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

salmeterol

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

N(CC(O)c2cc(c(cc2)O)Cc3c4c(c(c(c3)C(=O)O)O)cccc4)CCCCCCOCCCCc1ccccc1

InChI

1S/C36H43NO6/c38-33-18-17-27(22-29(33)23-28-24-32(36(41)42)35(40)31-16-7-6-15-30(28)31)34(39)25-37-19-9-1-2-10-20-43-21-11-8-14-26-12-4-3-5-13-26/h3-7,12-13,15-18,22,24,34,37-40H,1-2,8-11,14,19-21,23,25H2,(H,41,42)

InChI key

RQBBDIIWJKBGAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salmeterol impurity 1 BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Fluticasone and Salmeterol Inhalation Powder
  • Fluticasone and Salmeterol Inhalation Powder, pre-metered
  • Salmeterol Inhalation Powder, pre-metered
  • Packaging

    Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service