Skip to Content
Merck
All Photos(1)

Documents

843126

TOYOPEARL® Phenyl-650C Bulk Media

phase phenyl, bottle of 25 mL, 100 μm particle size

Synonym(s):

TOYOPEARL® Phenyl-650 Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TOYOPEARL®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 43126

parameter

≤50% organic solvent
3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

phenyl phase

particle size

100 μm

operating pH

1-13

capacity

40 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl Phenyl-650 resin is the middle hydrophobicity of the five HIC ligands offered by Tosoh Bioscience. Primary applications are for the separation of proteins, their isoforms, and aggregate removal. It is typically used in intermediate purification and polishing steps. For high performance separations, the use of TSKgel Phenyl-5PW resin is suggested. Both products have the same selectivity.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service