Skip to Content
Merck
All Photos(1)

Key Documents

822714

TSKgel® ODS-100V (5 µm) HPLC Columns

L x I.D. 2 cm × 3 mm, C18 (octadecyl) phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€372.00

€372.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€372.00

About This Item

UNSPSC Code:
41115711

€372.00


Please contact Customer Service for Availability

Product Name

TSKgel® ODS-100V HPLC Column, phase C18 (octadecyl), L × I.D. 2 cm × 3 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh 22714

extent of labeling

15% Carbon, endcapped with bifunctional dialkylsilane

parameter

0-100% organic solvent
0.43 mL/min standard flow rate
10-50 °C temperature
90 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

2 cm × 3 mm

surface area

450 m2/g

matrix

spherical silica

matrix active group

C18 (octadecyl) phase

particle size

5 μm

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® ODS-100V and TSKgel ODS-100Z columns incorporate the best-in-class surface properties to limit secondary interactions of basic, acidic and chelating compounds. Take advantage of the benefits of high column efficiency and symmetrical peak shapes to reduce your analysis and method development time! The ultra high purity Type B base silica contains negligible amounts of metal ion impurities. Combined with monomeric bonding chemistry, this silica makes the best general purpose Reversed Phase columns suitable for the most demanding separations in quality control as well as in research and development. TSKgel ODS-100V columns have a lower carbon content than ODS-100Z columns, making the bonded phase surface 100% water wettable and the preferred choice to retain polar metabolites.

Physical form

Shipped in acetonitrile.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service