Skip to Content
Merck
All Photos(1)

Key Documents

819804

TOYOPEARL® DEAE-650S Bulk Media

bottle of 25 mL, phase DEAE (diethylaminoethyl), 35 μm particle size

Synonym(s):

TOYOPEARL® DEAE-650 Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€190.00

€190.00


Please contact Customer Service for Availability


Select a Size

Change View
25 ML
€190.00

About This Item

UNSPSC Code:
41115700

€190.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 19804

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase

particle size

35 μm

operating pH

2-13

capacity

30 g/L±5 adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl DEAE-650 resin is a weak anion exchanger for blood fractionation and other biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps. If more powerful resolution is needed TSKgel DEAE-5PW resin should be used. Selectivity remains the same.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service