Skip to Content
Merck
All Photos(1)

Documents

817503

TSKgel® Octadecyl-2PW (5 µm) HPLC Columns

L x I.D. 1 cm × 6 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® Octadecyl-2PW Guard Column, phase C18 (octadecyl), L × I.D. 1 cm × 6 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh Octadecyl-2PW, 17503

extent of labeling

1 meq/mL ligand density (estimate)

parameter

0-100% organic solvent
4-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 6 mm

matrix

polymer particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

125 Å

operating pH

2-12

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Guard Column for 6 mmID TSKgel Octadecyl-2PW columns. TSKgel Guard Columns are stand-alone and do not require separate holders.

Physical form

Shipped in 55% acetonitrile - 45% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service