Skip to Content
Merck
All Photos(1)

Documents

814720

TSKgel® Phenyl-5PW (30) Bulk Media

bottle of 250 mL, phase phenyl, low Ligand density, 30 μm particle size

Synonym(s):

TSKgel® Phenyl-5PW HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product line

TSKgel®

form

slurry

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 14720

parameter

20 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

phenyl phase, low Ligand density

particle size

30 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Phenyl-5PW resin is the middle hydrophobicity of the five HIC ligands offered by Tosoh Bioscience. Primary applications are for the separation of proteins, their isoforms, and aggregate removal. It is typically used for high performance chromatography and polishing steps. It has the same selectivity as the larger particle Toyopearl Phenyl-650 products.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

103.1 °F

Flash Point(C)

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service