Skip to Content
Merck
All Photos(1)

Key Documents

807125

TSKgel® ODS-120T (5 µm) HPLC Columns

L x I.D. 25 cm × 4.6 mm, C18 (octadecyl) phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€775.00

€775.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€775.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

€775.00


Please contact Customer Service for Availability

Product Name

TSKgel® ODS-120T HPLC Column, phase C18 (octadecyl), L × I.D. 25 cm × 4.6 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh ODS-120T, 07125

extent of labeling

22% Carbon, multilayer, encapped

parameter

0-100% organic solvent
1.2 mL/min flow rate
10-50 °C temperature
210 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

200 m2/g

matrix

silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

150 Å pore diameter

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel ODS-120T columns use a 150Å pore size silica base support. The silica′s exclusion limit of 10,000 Da makes them a good choice for the reversed phase chromatography of peptides, small proteins, and small molecular weight compounds in organic and environmental samples. The columns are endcapped with trimethyl silane groups to improve the peak shape of negatively charged analytes. Bonded phase pore size is indicated by the number in the product description, in this case TSKgel ODS-120T has 120Å nominal pore size.

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service