Skip to Content
Merck
All Photos(1)

Key Documents

59482

Sapphire Piston

for use with 2010, Varian 00-997-261-08

Synonym(s):

Optimize Technologies HPLC Pump Part for Varian®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

packaging

pkg of 1 ea

manufacturer/tradename

Varian 00-997-261-08

compatibility

for use with 2010

Looking for similar products? Visit Product Comparison Guide

Legal Information

Varian is a registered trademark of Varian Medical Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service