Documents
59477
Optimize Technologies HPLC Pump Part for Spectra-Physics
for use with 8800, 8810, Isochrom, P-Series, Spectra Physics A3102-010
About This Item
UNSPSC Code:
41115854
packaging
pkg of 1
manufacturer/tradename
Spectra Physics A3102-010
compatibility
for use with 8800, 8810, Isochrom, P-Series
Features and Benefits
  • Offers optimal concentricity.        
  • Specifically cut in relation to the grain of the crystal lattice.         Maximal resistance to breakage.        
  • Superior wear characteristics.       
  • Proprietary attachment of the rod to the piston ferrule ensures concentricity.
Choose from one of the most recent versions:
Certificates of Analysis (COA)