Skip to Content
Merck
All Photos(1)

Documents

54797-U

Supelco

LiChrospher® Si-60 (5 µm) HPLC Columns

L × I.D. 1 cm × 4.6 mm, HPLC guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

LiChrospher® Si-60 HPLC Guard Cartridge, 5 μm particle size, L × I.D. 1 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L3

product line

LiChrospher®

feature

endcapped: no

packaging

pkg of 4 ea

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

surface area

700 m2/g

matrix

fully porous particle

matrix active group

silica phase

particle size

5 μm

pore size

60 Å

operating pH

2-7.5

application(s)

food and beverages

compatibility

for use with 3.0 to 4.6 mm column

separation technique

hydrophilic interaction (HILIC)
normal phase

General description

Use guard cartridge holder 54987

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service