Documents
509655
Syringes Fitted with Luer Lock Valve (509353)
volume 100 mL, 100MR-VLLMA-GT, needle size (not included)
About This Item
UNSPSC Code:
41122001
material
glass syringe
needle size
(not included)
feature
Pre-fitted with Luer Lock Valve
packaging
pkg of 1 ea
manufacturer/tradename
SGE 009770
volume
100 mL
Related Categories
General description
  • SGE Syringes Fitted with Luer Lock Valve, Model: 100MR-VLLMA-GT.
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Feature an interference fit between the plunger tip and the barrel, which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Choose from one of the most recent versions:
Certificates of Analysis (COA)