Skip to Content
Merck
All Photos(1)

Documents

48717-U

Supelco

Nitrobenzene-d5 solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal/surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

LQNUZADURLCDLV-RALIUCGRSA-N

General description

Nitrobenzene-d5 solution is an internal/surrogate standard specifically designed for monitoring organic chemicals on the Priority Pollutants List in municipal and industrial wastewater, per methods developed by the US EPA Environmental Monitoring Systems Laboratory in Cincinnati, Ohio (EMSL-CI), under the authority of the Clean Water Act (CWA).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service