Skip to Content
Merck
All Photos(1)

Key Documents

47661-U

Supelco

Methyl chloroacetate

certified reference material, 1000 μg/mL in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOCH3
CAS Number:
Molecular Weight:
108.52
Beilstein:
506255
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 552.2

vapor density

3.8 (vs air)

vapor pressure

10 mmHg ( 25 °C)
5.25 mmHg ( 20 °C)

CofA

certificate of analysis is enclosed in each package.

autoignition temp.

869 °F

feature

standard type calibration

expl. lim.

18.5 %

packaging

ampule of 1 mL

concentration

1000 μg/mL in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.422 (lit.)

bp

130 °C/740 mmHg (lit.)

mp

−33 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

application(s)

environmental

format

single component solution

storage temp.

room temp

SMILES string

COC(=O)CCl

InChI

1S/C3H5ClO2/c1-6-3(5)2-4/h2H2,1H3

InChI key

QABLOFMHHSOFRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniele Tedesco et al.
Chirality, 27(12), 914-918 (2015-10-09)
Caffeic acid-derived polyethers are a class of natural products isolated from the root extracts of comfrey and bugloss, which are endowed with intriguing pharmacological properties as anticancer agents. The synthesis of new polyether derivatives is achieved through ring-opening polymerization of
Milena E Miska et al.
Rapid communications in mass spectrometry : RCM, 29(24), 2341-2348 (2015-11-14)
The environmental occurrence of chlorinated acetic acids (CAAs) has been extensively studied, but the sources and transport are still not yet fully understood. A promising approach for source apportionment and process studies is the isotopic characterization of target compounds. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service