303400
Trichloroethylene in nitrogen
10 ppm, cylinder of 14 L, analytical standard
About This Item
size
Scotty 14
grade
analytical standard
form
gas
packaging
cylinder of 14 L
manufacturer/tradename
Scott Gas T257-14
availability
not available in Canada, EU
concentration
10 ppm
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
- Trichloroethylene in nitrogen, 10 ppm is a multi-component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors and in situ industrial stack analyzers.
- A certificate of analysis is included.
- Packaged in lightweight, non-refillable, recyclable containers.
- Portable, convenient, and easy to use.
- Manufactured gravimetrically for maximum accuracy.
- NIST traceable.
regulator
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service