29847-U
SGE Syringes for eVol® XCHANGE® Analytical Syringe Digital Drive
volume 0.2-5 μL, needle size 25 ga, needle L × O.D. × I.D. 50 mm × 0.5 mm × 0.2 mm, bevel tip stainless steel needle, pkg of 1 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle
needle size
25 ga
feature
5 μL eVol® syringe
packaging
pkg of 1 ea
manufacturer/tradename
SGE 2910020
needle L × O.D. × I.D.
50 mm × 0.5 mm × 0.2 mm
volume
0.2-5 μL
Looking for similar products? Visit Product Comparison Guide
General description
XCHANGE® analytical syringes can be easily and quickly changed. This allows the user to choose the best syringe for the volume being measured. It also allows the user to dedicate individual syringe to specific liquids or methods, reducing the possibility of cross-contamination. Only a few XCHANGE syringes are requied to dispense liquid volumes from 0.2 μL to 500 μL.
Legal Information
XCHANGE is a registered trademark of SGE Analytical Science Pty Ltd.
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service