Skip to Content
Merck
All Photos(1)

Key Documents

25054

Supelco

Tenax® GR

stainless steel TD tube, preconditioned

Synonym(s):

Tenax® GR Tube, stainless steel, for PerkinElmer® ATD, 10 pk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

material

stainless steel TD tube

feature

preconditioned

packaging

pkg of 10 tubes

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax GR

suitability

suitable for (extends range of Tenax TA)

application(s)

environmental
industrial hygiene

compatibility

for use with PerkinElmer ATD-50,ATD-400 and TurboMatrix (TMX) Series

Looking for similar products? Visit Product Comparison Guide

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.
Tenax is a registered trademark of Buchem B.V.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service