20262
ORBO™ 42 Small Supelpak™-20 specially treated Amberlite™ XAD®-2 (20/40), 66/33 mg
W,F,F (specially treated) separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Synonym(s):
ORBO™ Supelpak™ 20 series Tube
About This Item
Recommended Products
material
W,F,F (specially treated) separators
product line
Amberlite™
ORBO™
composition
Bed A, 66 mg
Bed B, 33 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 42 Small
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 75 mm
matrix
XAD-2 (specially treated)
particle size
20-40 mesh
application(s)
air monitoring
environmental
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service