Skip to Content
Merck
All Photos(1)

Documents

20124

Supelco

Chromosorb® W

AW, 80-100 mesh particle size, bottle of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

form

solid

packaging

bottle of 100 g

technique(s)

gas chromatography (GC): suitable

surface area

1.0-3.5 m2/g

particle size

80-100 mesh

density

0.21-0.27 g/mL at 25 °C
0.18 g/cm3 (loose weight)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromosorb® W is a diatomite which provides solid support for the packed GC columns. It is suitable for use with polar compounds, due to its low surface area and high inertness.

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guodong Sheng et al.
Environmental science and pollution research international, 20(6), 3708-3717 (2012-11-13)
This work determined the influence of humic acid (HA) and fulvic acid (FA) on the interaction mechanism and microstructure of Ni(II) onto diatomite by using batch experiments, X-ray photoelectron spectroscopy (XPS), and extended X-ray absorption fine structure (EXAFS) methods. Macroscopic
Baiyang Hu et al.
Journal of hazardous materials, 217-218, 85-91 (2012-04-03)
We developed a spongiform adsorbent that contains Prussian blue, which showed a high capacity for eliminating cesium. An in situ synthesizing approach was used to synthesize Prussian blue inside diatomite cavities. Highly dispersed carbon nanotubes (CNTs) were used to form
Maria Aivalioti et al.
Journal of hazardous materials, 207-208, 117-127 (2011-04-16)
The removal of BTEX (benzene, toluene, ethyl-benzene and m-,p-,o-xylenes), MTBE (methyl tertiary butyl ether) and TAME (tertiary amyl methyl ether) from aqueous solutions by raw, thermally, chemically and both chemically and thermally treated diatomite was studied, through batch adsorption experiments.
Norimasa Washizawa et al.
Dental materials journal, 31(4), 629-634 (2012-08-07)
The purpose of this study was to synthesize biomaterials from daily dental waste. Since alginate impression material contains silica and calcium salts, we aimed to synthesize calcium silicate cement from alginate impression material. Gypsum-based investment material was also investigated as
Junhua Li et al.
Applied microbiology and biotechnology, 97(6), 2541-2549 (2012-08-29)
The ribosomal protein L2, a constituent protein of the 50S large ribosomal subunit, can be used as Si-tag using silica particles for the immobilization and purification of recombinant proteins (Ikeda et al. (Protein Expr Purif 71:91-95, 2010); Taniguchi et al.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service