Skip to Content
Merck
All Photos(1)

Key Documents

13660

Supelco

Silica without Fluorescence

Grade 653XWP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
PubChem Substance ID:

grade

Grade 653XWP
high-purity grade (Davisil Grade 653XWP)

packaging

jar of 100 g

manufacturer/tradename

Davisil® 653XWP

particle size

230-400 mesh
35-70 μm

pore size

1.1-1.2 cm3/g pore volume
300 Å pore size

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Davisil is a registered trademark of W.R. Grace & Co.-Conn.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service