Skip to Content
Merck
All Photos(1)

Key Documents

13035-U

Supelco

Metal Packed GC Column (General Configuration)

phase 20% OV-101, matrix 80/100 Chromosorb W HP support, L × O.D. × I.D. 4.0 ft (1.2 m) × 1/8 in. × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
NACRES:
SB.54

material

stainless steel column

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

4.0 ft (1.2 m) × 1/8 in. × 2.1 mm

matrix

80/100 Chromosorb W HP support

matrix active group

20% OV-101 phase

compatibility

for use with Agilent Configuration A

General description

Packed columns the carrier gas percolates through the porous bed. In a packed column, separation is achieved by moving the solutes apart. Packed columns are made up of two main materials; stainless steel and Pyrex glass. Metal packed columns are more suitable for long and high carrier gas flow rates to tolerate the elevated pressures.[1] Chromosorb W, a fragile white solid prepared heating the diatomaceous earth with a flux. The resulting fine structure is cemented over glass which results in large and irregular pores.[2] HP is the classification of a support as high performance grade.[3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert L. Grob and Eugene F. Barry
Modern Practice of Gas Chromatography, 76-76 (2004)
Ian A. Fowlis
Gas Chromatography, 27-27 (2008)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service