Skip to Content
Merck
All Photos(4)

Key Documents

ZRB2808

Sigma-Aldrich

Anti-SALL4 Antibody, clone 1H18 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 112.23 kDa
observed mol wt ~160 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SALL4(57167)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H18 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects SALL4. It targets an epitope within 25 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal region of human SALL4.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from NTERA-2 cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected SALL4 in lysate from NTERA-2 cells.

Tested Applications

Western Blotting Analysis: A 1:50,000 dilution from a representative lot detected SALL4 in lysate from NCCIT cells and human testis and mouse testis tissues.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected SALL4 in Caco-2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected SALL4 in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Sal-like protein 4 (UniProt: Q9UJQ4; also known as Zinc finger protein 797, Zinc finger protein SALL4) is encoded by the SALL4 (also known as ZNF797) gene (Gene ID: 57167) in human. Sal-like protein 4 is a Transcription factor belongs to the spalt-like protein family with a key role in the maintenance and self-renewal of embryonic and hematopoietic stem cells. SALL4 is characterized with a multiple zinc finger (ZnF) domains including one N-terminal C2HC-type ZnF and seven C2H2-type ZnF domains. SALL4 is primarily localized in the nucleus, consistent with its function as a transcription factor. In vertebrates, its found in testis, thyroid gland and is abundantly expressed in both embryonic and adult stem/stem-like cells. The expression is tightly regulated, as inappropriate expression is associated with developmental abnormalities and cancer. It is suggested to play a role in the pathogenesis of leukemia stem cells and is reported to be constitutively expressed in human acute myeloid leukemia (AML) and induces AML in transgenic mice. It has some role in All-trans retinoic acid (ATRA)-based regimes in case of leukemia therapy. Additionally, aberrant expression of SALL4 is linked to various cancers, gastric cancer, hepatocellular cancer (HCC) and colorectal cancer (CRC), where it supports the proliferation and survival of cancer cells, often correlating with a poor prognosis. It may also act as a biomarker for HCC. SALL4 plays a crucial role in the maintenance of pluripotency and self-renewal in embryonic stem cells. It interacts with other key pluripotency factors, such as OCT4 and NANOG, forming regulatory networks essential for stem cell function. Defects in this SALL4 gene are a known cause of a rare genetic disorder Okihiro/Duane-radial ray syndrome (DRRS), which involves limb and heart defects, among other symptoms. The mutation or dysfunction of the SALL4 gene leads to this developmental syndrome, underscoring its importance in normal embryonic development. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Kohlhase, J., et al. (2005) Hum Mutat. Sep;26(3):176-83).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service