Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2590

Sigma-Aldrich

Anti-GDF15/NAG-1 Antibody, clone 1K11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

MIC-1, Macrophage inhibitory cytokine 1, NAG-1, NRG-1, NSAID-activated gene 1 protein, NSAID-regulated gene 1 protein, Placental TGF-beta, Placental bone morphogenetic protein, Prostate differentiation factor

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€188.00
4 X 25 μL
€386.00

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€188.00
4 X 25 μL
€386.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1K11
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 34 kDa
observed mol wt ~34 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... GDF15(9518)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1K11 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Growth/differentiation factor 15 (GDF15). It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Growth/differentiation factor 15 (GDF15).

Application

Quality Control Testing

Evaluated by Western Blotting in LNCap cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GDF15/NAG-1 in LNCap cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GDF15/NAG-1 in HT-1080 and NCI-H460 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound GDF15/NAG-1 peptide with a KD of 1.5 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected GDF15/NAG-1 in one million NCI-H460 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected GDF15/NAG-1 in HepG2 and HT1080 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GDF15/NAG-1 in Human placenta tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Growth/differentiation factor 15 (UniProt: Q99988; also known as GDF-15, Macrophage inhibitory cytokine 1, MIC-1, NSAID-activated gene 1 protein, NAG-1, NSAID-regulated gene 1 protein, NRG-1, Placental TGF-beta, Placental bone morphogenetic protein, and Prostate differentiation factor) is encoded by the GDF15 (also known as MIC1, PDF, PLAB, PTGFB) gene (Gene ID: 9518) in human. GDF15/NAG-1 is a disulfide-linked homodimeric protein that belongs to the TGF-β superfamily of cytokines. It is synthesized with a signal peptide (aa 1-29) and a propeptide (aa 30-194) that are subsequently cleaved off by furin like proprotein convertases to produce the mature active form. It is highly expressed in placenta and is detected at lower levels in prostate and colon tissue. Its expression is up-regulated in obesity. It is often induced under stress conditions, probably to maintain cell and tissue homeostasis. A moderate increase in serum GDF15 level observed with age. Highly elevated GDF15 levels are mostly linked to pathological conditions, including inflammation, myocardial ischemia, and cancer. It regulates food intake, energy expenditure, and body weight in response to metabolic and toxin-induced stress. It binds to its receptor GDNF family receptor alpha like (GFRAL) and activates GFRAL-expressing neurons localized in the area postrema and nucleus tractus solitarius of the brainstem. It then triggers the activation of neurons localized within the parabrachial nucleus and central amygdala, which constitutes part of the ′emergency circuit′ that shapes feeding responses to stressful conditions. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wischhusen, J., et al. (2020). Front. Immunol. 11; 951; Hsu, JY., et al. (2017). Nature. 550(7675); 255-259; Wang-Wei Tsai, V., et al. (2013). PLoS One. 8(2); e55174).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service