Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1825

Sigma-Aldrich

Anti-STAG2 Antibody, clone 2B22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€394.00

€192.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B22, recombinant monoclonal

description

2B22 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 141.33 kDa
observed mol wt ~140 kDa

purified by

using Protein A

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

canine, monkey, porcine, bovine, feline

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... STAG2(10735)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2B22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects STAG2. It targets an epitope within 24 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal region of human Stromal antigen 2 (STAG2).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected STAG2 in Jurkat cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected STAG2 in NIH3T3 and C6 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected STAG2 in MCF-7 cells.

Affinity Binding Assay: A representative lot of this antibody bound STAG2 with a KD of 8.7 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Stromal antigen 2 (UniProt: Q8N3U4; also known as SCC3 homolog 2, Cohesin subunit SA-2, STAG2) is encoded by the STAG2 (also known as SA2) gene (Gene ID: 10735) in human. STAG2 is a component of a an evolutionarily conserved, multimeric protein complex, known as the cohesion complex, which is required for the cohesion of sister chromatids after DNA replication. In human somatic cells, cohesin consists of four core subunits: SMC1, SMC3, SCC1, and one of two related helical repeat proteins known as STAG1 or STAG2. This complex apparently forms a large proteinaceous ring within which sister chromatids can be trapped. The cohesin complex is also reported to play a role in spindle pole assembly during mitosis. At anaphase, the RAD21 subunit of cohesion is cleaved, which leads the dissociation of the complex from chromatin and allows sister chromatids to segregate. During prophase, most of cohesin complex is shown to dissociate from chromatin probably because of phosphorylation by PLK1. However, the complex remains intact at centromeres. In the cohesion complex, STAG2 gene exhibits high mutation rate with about 90% mutations resulting in the introduction of premature codons that leads to loss of protein function. STAG2 mutations are known to cause Mullegama-Klein-Martinez syndrome, a neurodevelopmental disorder leading to intellectual deficiency, hearing loss, developmental delay, and congenital heart defect. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Smith, JS., et al. (2020). J. Transl. Med.18; Article number: 339; Soardi, FC., et al. (2017). Genom. Med. 2; Article 7).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service