Skip to Content
Merck
All Photos(8)

Key Documents

ZRB1822

Sigma-Aldrich

Anti-RSK-2 Antibody, clone 3O19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

90 kDa ribosomal protein S6 kinase 3, EC:2.7.11.1, ISPK-1, Insulin-stimulated protein kinase 1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, Ribosomal S6 kinase 2, Ribosomal protein S6 kinase alpha-3, S6K-alpha-3, p90-RSK 3, p90RSK3, pp90RSK2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€188.00
4 X 25 μL
€386.00

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€188.00
4 X 25 μL
€386.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€188.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3O19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

monkey, rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3O19 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Ribosomal S6 kinase 2 (RSK-2). It targets an epitope within 18 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human Ribosomal S6 kinase 2 (RSK-2).

Application

Quality Control Testing
Quality Control Testing


Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected RSK-2 in HepG2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected RSK-2 in A431 and COS-7 cell lysates and 1:1,000 dilution in NIH3T3 and C6 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RSK-2 in HeLa cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected RSK-2 in Human stomach and Human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Ribosomal protein S6 kinase alpha-3 (UniProt: P51812; also known as EC:2.7.11.1, S6K-alpha-3, 90 kDa ribosomal protein S6 kinase 3, p90-RSK 3, p90RSK3, Insulin-stimulated protein kinase 1, ISPK-1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, Ribosomal S6 kinase 2, RSK-2, pp90RSK2) is encoded by the RPS6KA3 (also known as ISPK1, MAPKAPK1B, RSK2) gene (Gene ID: 6197) in human. RSK-2 is a serine-threonine protein kinase that is expressed in various tissues with highest levels observed in the skeletal muscle. It acts downstream of ERK2/ERK1 signaling and mediates mitogenic and stress-induced activation of various transcription factors, including CREB1, ETV1/ER81 and NUR77, and regulates translation through RPS6 and EIF4B phosphorylation. It mediates cell proliferation, survival, and differentiation by modulating mTOR signaling and repressing pro-apoptotic function of BAD and death-associated protein kinase 1 (DAPK1). Upon insulin stimulation, it acts indirectly on the transcription regulation of several genes by phosphorylating GSK-3β at serine 9 and inhibiting its activity. RSK-2 is also involved in the mTOR nutrient-sensing pathway by directly phosphorylating TSC2 at serine 1798 that inhibits TSC2 ability to suppress mTOR signaling. It also mediates phosphorylation of RPTOR, that regulates mTORC1 activity and may promote rapamycin-sensitive signaling independently of the PI3K/AKT pathway. RSK-2 is activated by phosphorylation at serine 227 by 3-phosphoinositide dependent protein kinase 1 (PDPK1). This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Carrière, A., et a. (2008). Curr. Biol. 18(17); 1269-1277; Anjum, R., et al. (2005). Curr. Biol. 15(19); 1762-1767; Sutherland, C., et al. (1993). Biochem. J. 296(Pt1); 15-19).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service