Key Documents
Z692441
Cellometer® automated cell counting system
Auto T4 Plus, AC/DC input 100 / 240 V AC
Synonym(s):
Hemacytometer, automated cell counters, cell analysis, cell counting
About This Item
UNSPSC Code:
41161502
feature
CE compliant
packaging
pkg of 1 kit
manufacturer/tradename
Nexcelom Cellometer Auto T4 Plus
availability
not available in EU (available only in Belgum, Luxemburg, Netherlands, Spain, and Portugal)
General description
Cellometer Auto T4 offers simple-to-use software and user interface, and takes up almost no bench space. Because it works with your desktop computer or laptop, you get a full screen display, easy data input, and the ability to import/export data to a local or network location. The ideal choice for counting cell lines, Cellometer Auto T4 is easily shared among multiple users. Each user can customize cell types for improved accuracy and store them in a drop-down menu for easy access.
Cellometer automated cell counters eliminate manual cell counting and deliver more accurate, consistent results in a fraction of the time. Typically in less than 30 seconds per sample, Cellometer cell counters provide cell sample analysis results that include cell count, concentration, diameter, images, and trypan blue viability.
With the disposable counting chambers, there is no washing between samples, no risk of broken glass, and no sample cross-contamination. It can accurately recognize 300+ cell types from 2 to 250 microns in size, including cells of irregular shape, cells in clumps, cells with large size variation, and in samples with high levels of debris.
The innovative imaging and fluorescence technology of the Cellometer allows you to:
  • View cell morphology in real time, for visual confirmation after cell counting
  • Take advantage of 300+ cell types and easy, wizard-based parameter set-up
  • Save sample images with results securely on your computer, plus autosave results on the network for added convenience and data protection
Legal Information
Cellometer is a registered trademark of Nexcelom Bioscience, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)