Z659614
Bandelin SONOREX™ Ultrasonic bath accessories
insert basket for Z659592
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
material
stainless steel
manufacturer/tradename
Bandelin K 3 CL
availability
not available in USA and Canada
L × W × D
200 mm × 113 mm × 40 mm
General description
During ultrasonic operation, nothing may be placed on the tank floor, as this could cause abrasion between the sonication material and the ultrasonic oscillating tank. The use of a suitable basket to hold the sonication material is therefore recommended. This stainless steel baskets is designed so that the sonication material is always in the optimal area of the cavitation field (1-2 cm from the floor) in the oscillating tank. With the help of a basket, the sonication material can also be conveniently placed in the device and removed after operation, e.g. for rinsing, without the user coming into contact with the sonication liquid in the oscillating tank.
Features and Benefits
- perfect positioning in the sound field
- optimal loading and unloading
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)