Skip to Content
Merck
All Photos(1)

Key Documents

Z637238

Edisonite universal detergent

weight 5 kg

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
€136.00

€136.00


Please contact Customer Service for Availability


Select a Size

Change View
5 KG
€136.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

€136.00


Please contact Customer Service for Availability

description

pulverized detergents

manufacturer/tradename

BRAND 44205

technique(s)

protein quantification: suitable

weight

5 kg

Looking for similar products? Visit Product Comparison Guide

General description

Pulverized detergents for immersion and ultrasonic baths. Alkaline. Ideal for removal of natural fats and oils, proteins, and blood.
Ingredients: Special phosphates, surfactants, emulsifying agents, auxiliary agents.

Other Notes

BR44205 (India)

Legal Information

Edisonite is a trademark of Merz GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service