Skip to Content
Merck
All Photos(1)

Key Documents

Z378496

Sigma-Aldrich

Cell Biology: A Laboratory Handbook, 2nd ed.

Sign Into View Organizational & Contract Pricing


About This Item

ISBN-10:
0-12-164725-0
ISBN-13:
978-0-12-164725-4
UNSPSC Code:
55101509

publication info

J.E. Celis, ed., Academic Press, 1997, 2400 pp., comb bound

manufacturer/tradename

Elsevier

General description

A superb handbook, this four-volume set should be a required resource for any biology or biochemistry research facility. Contributors are world-renowned in their fields, and the content has been peer-reviewed. The 260 chapters are uniformly edited and beautifully illustrated; 40% of the material is new to this edition. As an encyclopedic manual of up-to-date instructions for cell biology techniques and transitions to molecular biology, it′s the next best thing to having the expert at your side.

Table of Contents

Volume 1
Part 1: Cell and Tissue Culture and Associated Techniques
Part 2: Viruses

Volume 2
Part 3: Organelles and Cellular Structures
Part 4: Assays
Part 5: Antibodies
Part 6: Immunocytochemistry
Part 7: Vital Staining of Cells

Volume 3
Part 8: Light Microscopy and Contrast Generation
Part 9: Electron Microscopy
Part 10: Intracellular Measurements
Part 11: Cytogenetics and in Situ Hybridization
Part 12: Transgenic and Gene Knockouts

Volume 4
Part 13: Transfer of Macromolecules and Small Molecules
Part 14: Expression Systems
Part 15: Differential Gene Expression
Part 16: Proteins

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service