Skip to Content
Merck
All Photos(1)

Documents

Z317047

Eppendorf® GELoader® tips

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121606
NACRES:
NB.01

sterility

non-sterile

packaging

pack of 192 2 racks of 96

manufacturer/tradename

Eppendorf® 0030 001.222 (EU)
Eppendorf® 022351656 (USA)

capillary section I.D. × L

0.3 mm × 15 mm

General description

Flexible tip specially designed for loading samples onto sequencing gels.

Application

  • Eppendorf® GELoader® tips have been used for packing of titanium dioxide microcolumns to purify sialic acid (SA)-containing glycopeptides.
  • In MALDI-FTMS (matrix-assisted laser desorption/ionization-Fourier transform mass spectrometer), they have been used for the deposition of matrix solution onto probe tip and for the deposition of samples onto matrix layers.
  • They have been used for packing of titanium dioxide microcolumns to purify phosphorylated peptides. They have also been used for preparing chromatographic reversed-phase microcolumns to desalt and concentrate peptides.
  • They have been used for loading of urine samples into nanospray tip of LTQ Orbitrap XL for MS/MS (mass spectrometry) measurements.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
GELoader is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploring the sialiome using titanium dioxide chromatography and mass spectrometry.
Larsen MR, et al.
Molecular and Cellular Proteomics, 6, 1778-1787 (2007)
Metabolomic screening using ESI-FT MS identifies potential radiation-responsive molecules in mouse urine.
Iizuka D, et al.
Journal of Radiation Research (2016)
Highly selective enrichment of phosphorylated peptides from peptide mixtures using titanium dioxide microcolumns.
Larsen MR, et al.
Molecular and Cellular Proteomics, 4, 873-886 (2005)
A general method for precalculation of parameters for sustained off resonance irradiation/collision-induced dissociation.
Mirgorodskaya E, et al.
Journal of the American Society For Mass Spectrometry, 13, 318-324 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service