Skip to Content
Merck
All Photos(1)

Key Documents

WHAWB120204

QIAcard FTA Wash Buffer

Nucleic acid purification buffer, bottle of 500 mL

Synonym(s):

QIAcard FTA Wash Buffer (500 ml), Whatman FTA, Whatman FTA cards, Z719978

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€431.00

€431.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€431.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

€431.00


Please contact Customer Service for Availability

feature

Nucleic acid purification buffer

packaging

bottle of 500 mL

manufacturer/tradename

Qiagen WB120204

General description

QIAcard FTA Wash Buffer is used for purifying nucleic acids captured and stored on FTA cards. This versatile reagent is recommended for use with QIAcard FTA and Indicating FTA Card formats, and EasiCollect formats.

Features and Benefits

  • This reagent can be used to wash punches in the "punch-in" procedure for standard STR amplification.
  • QIAcard FTA Wash Buffer allows for excellent results by removing PCR inhibitors and contaminants that can lower PCR yields.
  • 500 ml for an increased number of reactions.

Other Notes

Field of Use : For internal research use only

Legal Information

FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Description of common problems that can occur during genomic DNA preparation

This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.

Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.

Protocols

This document shows intructions for EasiCollect™ device from Cytiva.

This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.

A number of options are available for sample collection, transport, archiving, and DNA purification.

Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service