Skip to Content
Merck
All Photos(1)

Key Documents

T5651

Sigma-Aldrich

Tetrodotoxin with Citrate Buffer

pH 4.8, powder

Synonym(s):

Fugu poison, Maculotoxin, TTX, Tarichatoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N3O8
CAS Number:
Molecular Weight:
319.27
Beilstein:
49176
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

availability

not available in USA

pH

4.8

solubility

H2O: soluble (stable at pH 4-5 if stored frozen)
strong acid or alkaline solutions: unstable (destroyed by boiling at pH 2.)

storage temp.

2-8°C

SMILES string

OC[C@@]1(O)[C@H]2OC3(O)OC1C4[C@@H](O)NC(=N)NC4([C@@H]2O)[C@@H]3O

InChI

1S/C11H17N3O8/c12-8-13-6(17)2-4-9(19,1-15)5-3(16)10(2,14-8)7(18)11(20,21-4)22-5/h2-7,15-20H,1H2,(H3,12,13,14)/t2?,3-,4?,5+,6-,7+,9+,10?,11?/m1/s1

InChI key

CFMYXEVWODSLAX-IXUBDLNJSA-N

Gene Information

Application

Tetrodotoxin has been used to study its effects on microglial functions in rats2. This toxin has also been used to inactivate lateral magnocellular nucleus of the nidopallium (LMAN) in juvenile zebra finch. This study reported that the toxin causes a decrease in sequence and acoustic variability which result in the production of a stereotyped song by the motor pathway3.

Biochem/physiol Actions

Reversible, selective blocker of Na+ channels; blocks propagation of impulses in excitable membranes. Used to characterize sodium channels in excitable membranes and to study the role of sodium channels in normal physiology and disease.

Packaging

Vial contains 1 mg of tetrodotoxin and approx. 5 mg citrate buffer, pH 4.8.

Preparation Note

The product is water soluble and stable at pH 4-5 if it is stored frozen. It is unstable in strong acid and alkaline solutions and is rapidly destroyed by boiling at pH 2 and below.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas C Jaramillo et al.
Autism research : official journal of the International Society for Autism Research, 10(1), 42-65 (2016-08-06)
Mutations/deletions in the SHANK3 gene are associated with autism spectrum disorders and intellectual disability. Here, we present electrophysiological and behavioral consequences in novel heterozygous and homozygous mice with a transcriptional stop cassette inserted upstream of the PDZ domain-coding exons in
Scott R Burks et al.
Theranostics, 9(19), 5517-5531 (2019-09-20)
Pulsed focused ultrasound (pFUS) technology is being developed for clinical neuro/immune modulation and regenerative medicine. Biological signal transduction of pFUS forces can require mechanosensitive or voltage-gated plasma membrane ion channels. Previous studies suggested pFUS is capable of activating either channel
Hao Yang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 27(8), 1467-1482 (2019-06-04)
Irreversible neuron loss following spinal cord injury (SCI) usually results in persistent neurological dysfunction. The generation of autologous neural stem cells (NSCs) holds great potential for neural replenishment therapies and drug screening in SCI. Our recent studies demonstrated that mature
Miriam Michel et al.
Scientific reports, 9(1), 11888-11888 (2019-08-17)
Assessment of left ventricular mass (LVM) is important in the evaluation of patients with congenital heart disease (CHD) and cardiac magnetic resonance imaging (CMR) is the gold standard. Recent software allows LVM calculation by real-time 3-dimensional echocardiography (RT3DE). We investigated
Kun-Wei Wu et al.
Frontiers in cellular neuroscience, 11, 290-290 (2017-10-03)
Brain microvascular endothelial cells (BMEC) have been found to guide the migration, promote the survival and regulate the differentiation of neural cells. However, whether BMEC promote development and maturation of immature neurons is still unknown. Therefore, in this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service