Skip to Content
Merck
All Photos(1)

Key Documents

T3699

Sigma-Aldrich

Monoclonal Anti-T7 tag, Peroxidase conjugate antibody produced in mouse

2.0-3.0 mg/mL, purified immunoglobulin, lyophilized powder

Synonym(s):

Monoclonal Anti-T7 tag antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

biological source

mouse

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

T7 Tag

form

lyophilized powder

concentration

2.0-3.0 mg/mL

technique(s)

indirect ELISA: suitable

isotype

IgM

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Immunogen

synthetic peptide corresponding to the T7-tag (MASMTGGQQMG-K), conjugated to KLH

Physical form

Supplied as a lyophilized powder. After reconstitution the solution contains 1% BSA and 0.01% thimerosal in 0.01 M sodium phosphate buffered saline, pH 7.4.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria A Hassebroek et al.
Molecular biology of the cell, 31(23), 2537-2556 (2020-09-03)
Proper chromosome segregation is essential for faithful cell division and if not maintained results in defective cell function caused by the abnormal distribution of genetic information. Polo-like kinase 1-interacting checkpoint helicase (PICH) is a DNA translocase essential for chromosome bridge

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service