Skip to Content
Merck
All Photos(1)

Key Documents

T3317

Eppendorf® Thermomixer® R, dry block heating and cooling shaker

basic device without exchangeable thermoblock, accepts exchangeable thermoblocks, 120 V, 60 Hz, 1/cs

Synonym(s):

cooling mixer, cooling shaker, heating mixer, heating shaker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

AC/DC input

115 V AC

packaging

case of 1
pack of 1

manufacturer/tradename

Eppendorf® 022670107 (USA)

availability

not available in EU

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® Thermomixer R Dry Block Heating and Cooling Shaker

The Thermomixer R conveniently offers simultaneous mixing and temperature control. It meets even the highest demands for user-friendliness and flexibility: its′ counter-cooling ability provides efficient cooling, thereby greatly expanding both its temperature control functions and range of applications.

Ten different thermoblocks are available, all of which can be simply and quickly exchanged, to adapt the unit to daily and/or changing requirements in the laboratory. Two freely selectable program sequences with incubating, mixing and rest phases, as well as subsequent cooling to 13°C below room temperature, make everyday lab routines much easier.

The Eppendorf Thermomixer R combines shaking, heating, and cooling for maximum application versatility. Simultaneous mixing and incubation accelerate reaction processes and ease user′s workload. Choice of four exchangeable blocks provides flexibility for various vessels, accommodating 24 micro test tubes for 0.5, 1.5, and 2.0 mL volumes, as well as a 96-well or other microplates (sold separately).

  • Adjustable mixing speed for gentle to vigorous shaking
  • Autoclavable and freezer-safe numbered racks for quick, easy processing and transport
  • Short-mix function, ideal for quick "vortex" applications
  • Digital display of all parameters including actual and set temperature
  • Exchangeable blocks accommodate micro test tubes or microplates
  • Quick heat-up and cooling times for increased throughput
  • Two programs allow parameters to be stored for common or successive applications
  • Intermittent mixing may be programmed for applications like enzyme reactions or DNA hybridizations
  • Automatically recognizes Thermoblocks and displays parameters
  • RS-232 interface allows documentation of operating parameters.
  • Temperature range from 13°C below room temperature up to 99°C
  • Temperature Accuracy: 20°C to 45°C, ±0.5°C; below ambient and above 45 °C, ±2°C
  • Heating rate: approx. 5°C/minute
  • Mixing Speed: 300 to 1,500 rpm
  • Mixing Stroke: 3 mm
  • Dimensions, without block (W x D x H): 8.7 x 9.8 x 4.9 in. / 22 x 25 x 12.5 cm
  • Timer: 1 minute to 99 hours 59 minutes, or continuous

Note: Basic unit supplied without exchangeable thermoblock.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Thermomixer is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service