Skip to Content
Merck
All Photos(2)

Documents

T0252

Sigma-Aldrich

Tricholine citrate

≥65 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H14NO)3 · C6H5O7
CAS Number:
Molecular Weight:
501.61
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

concentration

≥65 wt. % in H2O

SMILES string

C[N+](C)(C)CCO.C[N+](C)(C)CCO.C[N+](C)(C)CCO.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3C5H14NO/c7-3(8)1-6(13,5(11)12)2-4(9)10;3*1-6(2,3)4-5-7/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*7H,4-5H2,1-3H3/q;3*+1/p-3

InChI key

RPERJPYDELTDMR-UHFFFAOYSA-K

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ha Yeon Sung et al.
Nature communications, 8(1), 1484-1484 (2017-11-15)
Animals must detect aversive compounds to survive. Bitter taste neurons express heterogeneous combinations of bitter receptors that diversify their response profiles, but this remains poorly understood. Here we describe groups of taste neurons in Drosophila that detect the same bitter
Bhanu Shrestha et al.
Insect biochemistry and molecular biology, 131, 103550-103550 (2021-02-08)
DEET is the most widely used active ingredient in insect repellents and offers protection against insect bites. We previously reported that DEET suppresses the feeding behavior of Drosophila, which is guided by gustatory receptors (GRs) in bitter-sensing gustatory receptor neurons.
Christina E May et al.
Cell reports, 27(6), 1675-1685 (2019-05-09)
Recent studies find that sugar tastes less intense to humans with obesity, but whether this sensory change is a cause or a consequence of obesity is unclear. To tackle this question, we study the effects of a high sugar diet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service