Skip to Content
Merck
All Photos(1)

Key Documents

SYNP160005D

Synthware cylindrical pressure vessell with protective coating

bottle capacity 100 mL, O.D. × L 46 mm × 110 mm, bushing type, #15

Synonym(s):

Synthware P160005D, pressure tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

feature

bushing type #15

packaging

pack of 1 ea

manufacturer/tradename

Kemtech P160005D

O.D. × L

46 mm × 110 mm

bottle capacity

100 mL

General description

Pressure Vessel, Heavy Wall, Cylindrical,100mL, #15, 46X110mm, Protective Covering, Working Temp -40°C To 80°C,Includes Accessories: B100015 + R111Ep

Application

kemetch; synthware; pressure vessel; cylindrical

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service