Skip to Content
Merck
All Photos(1)

Key Documents

SRP6383

Sigma-Aldrich

CD319/SLAMF7/CRACC human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

19A, CS1, FOAP-12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 23.2 kDa
observed mol wt 33-48 kDa (DTT-reduced. Protein migrates due to glycosylation. Ser 23 is the predicted N-terminus.)

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SLAMF7(57823)

General description

Research area: IMMUNO AND CKS

SLAM family member 7 (SLAMF7) is also known as CD2-like receptor-activating cytotoxic cells (CRACC), membrane protein FOAP-12, CD antigen CD319, Novel Ly9, Protein 19A, which is a single-pass type I membrane protein and a member of the CD2 family of cell surface receptors. SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. SLAM F7 (signaling lymphocytic activation molecule F7) receptors are expressed on immune cells such as activated T cells, most B cells, antibody-producing plasma cells and natural killer cells and also on myeloid cells. The SLAMF7 gene is mapped to human chromosome 1q23.3.

Biochem/physiol Actions

SLAMF7 (signaling lymphocytic activation molecule F7) is considered as a self-ligand which identifies another SLAMF7 expressed on the cell. Apart from immune cells, SLAMF7 is highly expressed in multiple myeloma cells.Isoform 1 of SLAMF7 mediates natural killer (NK) cell activation through a SH2 domain containing 1A (SH2D1A)-independent extracellular signal-regulated extracellular signal-regulated kinases (ERK) mediated pathway. It may play a role in lymphocyte adhesion.Isoform 3 of SLAMF7 does not mediate any NK cell activation.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immune cell inhibition by SLAMF7 is mediated by mechanism requiring Src kinases, CD45 and SHIP-1 defective in multiple myeloma cells.
Guo H, et al.
Molecular and Cellular Biology (2014)
Controlling natural killer cell responses: integration of signals for activation and inhibition.
Long E O, et al.
Annual Review of Immunology, 31, 227-258 (2013)
Elotuzumab and daratumumab: emerging new monoclonal antibodies for multiple myeloma.
Kuroda J, et al.
Expert Review of Anticancer Therapy, 13(9), 1081-1088 (2013)
SLAM receptors and SAP influence lymphocyte interactions, development and function.
Schwartzberg P L, et al.
Nature Reviews: Immunology, 9(1), 39-39 (2009)
Epigenetic suppression of the immunoregulator MZB1 is associated with the malignant phenotype of gastric cancer.
Kanda M, et al.
International Journal of Cancer. Journal International Du Cancer, 139(10), 2290-2298 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service