Skip to Content
Merck
All Photos(1)

Key Documents

SRP5108

Sigma-Aldrich

p21CIP1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAP20, CDKN1, CDKN1A, MDA-6, P21, SDI1, WAF1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€236.00

€236.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€236.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

€236.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~46 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDKN1A(1026)

General description

CIP 1 (Cyclin-Dependent Kinase Inhibitor 1A) regulates cell cycle progression, terminal differentiation, and apoptosis. CIP1 was shown to be induced by p53 and to be a potent inhibitor of cyclin-dependent kinase (CDK) activity. DNA damage leads to increased expression of CIP1 in cyclin E-containing complexes and to an associated decrease in cyclin-dependent kinase activity. CIP1 is a critical downstream effector in the p53-specific pathway of growth control in mammalian cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Wagner et al.
Anticancer research, 21(1A), 333-345 (2001-04-13)
The cdknlA gene encodes CDKN1A, a protein that regulates cell cycle progression, terminal differentiation, and apoptosis. Polymorphisms or loss of heterozygosity of this usually biallelically expressed gene have no major impact on carcinogenesis. The prevalence of somatic mutations in malignancies
W S el-Deiry et al.
Cancer research, 54(5), 1169-1174 (1994-03-01)
The tumor growth suppressor WAF1/CIP1 was recently shown to be induced by p53 and to be a potent inhibitor of cyclin-dependent kinases. In the present studies, we sought to determine the relationship between the expression of WAF1/CIP1 and endogenous regulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service