Skip to Content
Merck
All Photos(2)

Key Documents

SRP5092

Sigma-Aldrich

TIE2 (771-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CD202B, TEK, VMCM, VMCM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

10-14 nmol/min·mg

mol wt

~61 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TEK(7010)

General description

TIE 2 or TEK is a receptor tyrosine kinase that is expressed principally on vascular endothelium. Disrupting TIE 2 function in mice results in embryonic lethality with defects in embryonic vasculature, suggesting a role in blood vessel maturation and maintenance. Angiopoietin-1 is a secreted growth factor that binds to and activates the TIE 2 receptor tyrosine kinase. SHP2 and GRB2 are recruited to the activated TIE 2 kinase domain and are part of the cellular responses that mediate TIE 2 function. TIE 2 expression is upregulated in the endothelium of vascular "hot spots" in human breast cancer specimens. However, TIE 2 is also overexpressed in areas of active angiogenesis in normal tissues.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Woolf et al.
Pediatric nephrology (Berlin, Germany), 16(2), 177-184 (2001-03-23)
Angiopoietin-1 (Ang-1) is a secreted growth factor which binds to and activates the Tie-2 receptor tyrosine kinase. The factor enhances endothelial cell survival and capillary morphogenesis, and also limits capillary permeability. Ang-2 binds the same receptor but fails to activate
Kevin G Peters et al.
Recent progress in hormone research, 59, 51-71 (2004-01-30)
Abundant data now demonstrate that the growth of new blood vessels, termed angiogenesis, plays both pathological and beneficial roles in human disease. Based on these data, a tremendous effort has been undertaken to understand the molecular mechanisms that drive blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service