Skip to Content
Merck
All Photos(2)

Key Documents

SRP5077

Sigma-Aldrich

PTPN7, active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

BPTP-4, HEPTP, LPTP, PTPNI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1309-1771 nmol/min·mg

mol wt

~67 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN7(5778)

General description

PTPN7 gene is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The noncatalytic N-terminus of this PTP can interact with MAP kinases and negatively regulates ERK2 and p38 MAP-kinases activity. The PTPN7 was shown to be involved in the regulation of T cell antigen receptor (TCR) signaling, which was thought to function through dephosphorylating the molecules related to MAP kinase pathway.

Physical form

Supplied in 20mM MOPS, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25mM DTT, 0.1mM PMSF, 30% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Oh-hora et al.
Journal of immunology (Baltimore, Md. : 1950), 163(3), 1282-1288 (1999-07-22)
Leukocyte protein tyrosine phosphatase (LC-PTP)/hemopoietic PTP is a human cytoplasmic PTP that is predominantly expressed in the hemopoietic cells. Recently, it was reported that hemopoietic PTP inhibited TCR-mediated signal transduction. However, the precise mechanism of the inhibition was not identified.
M Adachi et al.
Cancer research, 52(3), 737-740 (1992-02-01)
Protein-tyrosine phosphatase (PTP)-related complementary DNAs from NALM-6 (pre-B cell line) were amplified by reverse transcriptase polymerase chain reaction using primers corresponding to the conserved catalytic domains of PTPs. Thirty-three polymerase chain reaction products, identified as PTP related complementary DNAs, were
S M Pettiford et al.
Oncogene, 19(7), 858-869 (2000-03-07)
HePTP is a tyrosine specific protein phosphatase that is strongly expressed in activated T-cells. It was recently demonstrated that in transfected T-cells HePTP impairs TCR-mediated activation of the MAP-kinase family members ERK2 and p38 and it was suggested that both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service