Skip to Content
Merck
All Photos(1)

Documents

SRP4639

Sigma-Aldrich

BMP-13 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

CDMP-2, Cartilage-Derived Morphogenetic Protein-2, GDF-6, bone morphogenetic protein 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized

potency

0.5-4 μg/mL ED50 (determined by its ability to induce alkaline phosphatase production by ATDC-5 chondrogenic cells.)

mol wt

~27 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDF6(392255)

General description

BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. These ligands mediate numerous physiological processes. BMPs initiate, promote and maintain cartilage and bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Expression of BMP-13 has been found in the hypertrophic chondrocytes of ossifying long bone centers. Recombinant human BMP-13/CDMP-2 is a 27 kDa homodimeric disulfide-linked protein consisting of two 120 amino acids polypeptide chains.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute to 0.1-1.0 mg/mL in water containing BSA (50 μg BSA per 1 μg of protein).

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service