Skip to Content
Merck
All Photos(1)

Documents

SRP3277

Sigma-Aldrich

SDF-1 BETA human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CXCL12, PBSF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20.0-80.0 ng/mL

mol wt

8.5 kDa

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL12(6387)

General description

SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV suppressive activity in cells expressing the CXCR4 receptor.

Biochem/physiol Actions

SDF-1α and β are stromal derived CXC chemokines, and signal through the CXCR4 receptor. Recombinant human SDF-1α is an 8.0 kDa protein containing 68 amino acid residues. Recombinant human SDF-1β is an 8.5 kDa protein containing 72 amino acid residues.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D'Apuzzo et al.
European journal of immunology, 27(7), 1788-1793 (1997-07-01)
In the bone marrow, progenitor (pro-) and precursor (pre-) B cells depend on close contact with stromal cells for growth and maturation. Stromal cell-derived factor 1 (SDF-1), also known as pre-B cell growth-stimulating factor, is produced by bone marrow stromal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service