Skip to Content
Merck
All Photos(1)

Documents

SRP3152

Sigma-Aldrich

SCGF-alpha human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Stem Cell Growth Factor-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤2.5 ng/mL ED50

mol wt

33.9 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CLEC11A(6320)

General description

SCGF alpha and beta are hematopoietic growth factor that exerts their activity at early stages of hematopoiesis. The SCGFs are non-glycosylated species-specific cytokines that can support growth of primitive hematopoietic cells, and in combination with EPO or GM-CSF, promote proliferation or erthroid or myeloid progenitors, respectively. Recombinant human SCGF-a is a 33.9 kDa polypeptide containing 305 amino acid residues.

Biochem/physiol Actions

SCGF alpha and beta are hematopoietic growth factor that exerts their activity at early stages of hematopoiesis. Recombinant human SCGF-a is a 33.9 kDa polypeptide containing 305 amino acid residues.

Sequence

GHGARGAERE WEGGWGGAQE EEREREALML KHLQEALGLP AGRGDENPAG TVEGKEDWEM EEDQGEEEEE EATPTPSSGP SPSPTPEDIV TYILGRLAGL DAGLHQLHVR LHALDTRVVE LTQGLRQLRN AAGDTRDAVQ ALQEAQGRAE REHGRLEGCL KGLRLGHKCF LLSRDFEAQA AAQARCTARG GSLAQPADRQ QMEALTRYLR AALAPYNWPV WLGVHDRRAE GLYLFENGQR VSFFAWHRSP RPELGAQPSA SPHPLSPDQP NGGTLENCVA QASDDGSWWD HDCQRRLYYV CEFPF

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Ding et al.
Fish & shellfish immunology, 33(3), 494-503 (2012-06-12)
The toll-like receptors (TLRs) are an important gene family in host innate immunologic surveillance. The TLR22 gene is an essential member of the TLRs that is only found in aquatic animals and has been detected in some bony fish. Here

Articles

Read article on hematopoietic cytokines and hematopoiesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service