Skip to Content
Merck
All Photos(1)

Key Documents

SRP3086

Sigma-Aldrich

IL-2 Receptor human

recombinant, expressed in (BTI-Tn-5B1-4) High-5 Insect Cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CD25 antigen, TAC-antigen, soluble IL-2 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in (BTI-Tn-5B1-4) High-5 Insect Cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.5-15 μg/mL ED50

mol wt

31 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL2RA(3559)

General description

The IL-2 receptor system consists of three non-covalently linked subunits termed IL-2Rα, IL-2Rβ, and IL-2Rγ. The IL-2Ra is a type I transmembrane protein consisting of a 219 amino acid extracellular domain, a 19 amino acid transmembrane domain and a 13 amino acid intracellular domain, which is not involved in the transduction of IL-2 signals. Proteolytic processing of IL-2Ra releases the entire extracellular domain of IL-2Ra thereby generating a 219 amino acid soluble protein called soluble IL-2Ra (sIL-2Ra). The homodimeric form binds IL-2 (KD=10 mM) and facilitates IL-2 signaling. The secreted sIL-2Ra is expressed on leukemia cells, lymphoma cells, newly activated T and B cells, as well as on approximately 10% of NK cells. Recombinant human sIL-2R alpha is a 24.8 kDa protein containing 219 amino acid residues consisting of only the extracellular domain of IL-2R alpha. Due to glycosylation, IL-2R alpha has an approximate molecular weight of 31 kDa based on SDS-PAGE gel and Mass Spectrometry.

Biochem/physiol Actions

The IL-2 receptor system consists of three non-covalently linked subunits termed IL-2Ra, IL-2Rb, and IL-2Rg. Recombinant human sIL-2R alpha is a 24.8 kDa protein containing 219 amino acid residues consisting of only the extracellular domain of IL-2R alpha. Due to glycosylation, IL-2R alpha has an approximate molecular weight of 31 kDa based on SDS-PAGE gel and Mass Spectrometry.

Sequence

ELCDDDPPEI PHATFKAMAY KEGTMLNCEC KRGFRRIKSG SLYMLCTGNS SHSSWDNQCQ CTSSATRNTT KQVTPQPEEQ KERKTTEMQS PMQPVDQASL PGHCREPPPW ENEATERIYH FVVGQMVYYQ CVQGYRALHR GPAESVCKMT HGKTRWTQPQ LICTGEMETS QFPGEEKPQA SPEGRPESET SCLVTTTDFQ IQTEMAATME TSIFTTEYQ

Physical form

Lyophilized from 1x PBS.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 1x PBS pH 7.2 to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smith, K.A., et al.
Cytokine Reference, 113-125 (2001)
K A Smith
Science (New York, N.Y.), 240(4856), 1169-1176 (1988-05-27)
Interleukin-2 (IL-2), the first of a series of lymphocytotrophic hormones to be recognized and completely characterized, is pivotal for the generation and regulation of the immune response. A T lymphocyte product, IL-2 also stimulates T cells to undergo cell cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service