Skip to Content
Merck
All Photos(2)

Documents

SRP0445

Sigma-Aldrich

Ataxin3 active human

recombinant, expressed in E. coli, ≥59% (SDS-PAGE)

Synonym(s):

AT3, ATX3, JOS, MJD, SCA3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥59% (SDS-PAGE)

form

aqueous solution

mol wt

42 kDa

packaging

pkg of 250 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ATXN3(4287)

General description

Ataxin3 (ATXN3) is encoded by the gene mapped to human chromosome 14q32. The encoded protein is characterized with a ubiquitin interaction motif (UIM) domain involved in binding mono and/or polyubiquitylated proteins.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Ataxin3 (ATXN3) is a polyglutamine neurodegenerative disease protein involved in the ubiquitin-proteasome pathway. ATXN3 performs all the functions same as ubiquitin proteases such as, reducing polyubiquitylation of 125I-lysozyme by removing ubiquitin from polyubiquitin chains, cleaving a ubiquitin protease substrate and binding the specific ubiquitin protease inhibitor, ubiquitin-aldehyde. Mutation in the gene has been observed in spinocerebellar ataxia type 3 patients.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ataxin-3 is a histone-binding protein with two independent transcriptional corepressor activities.
Li F, et al.
The Journal of Biological Chemistry, 277(47), 45004-45012 (2002)
Familial hemiplegic migraine: linkage to chromosome 14q32 in a Spanish kindred.
Cuenca-Leon E, et al.
Neurogenetics, 10(3), 191-198 (2009)
The polyglutamine neurodegenerative protein ataxin-3 binds polyubiquitylated proteins and has ubiquitin protease activity.
Burnett B, et al.
Human Molecular Genetics, 12(23), 3195-3205 (2003)
Maria do Carmo Costa et al.
PloS one, 5(7), e11728-e11728 (2010-07-30)
During myogenesis several transcription factors and regulators of protein synthesis and assembly are rapidly degraded by the ubiquitin-proteasome system (UPS). Given the potential role of the deubiquitinating enzyme (DUB) ataxin-3 in the UPS, and the high expression of the murine
Sandra Macedo-Ribeiro et al.
PloS one, 4(6), e5834-e5834 (2009-06-09)
Spinocerebellar ataxia type-3, also known as Machado-Joseph Disease (MJD), is one of many inherited neurodegenerative disorders caused by polyglutamine-encoding CAG repeat expansions in otherwise unrelated genes. Disease protein misfolding and aggregation, often within the nucleus of affected neurons, characterize polyglutamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service