Skip to Content
Merck
All Photos(1)

Documents

SML3822

Sigma-Aldrich

SA-15-P

≥98% (HPLC)

Synonym(s):

N-[4-[2-[2,5-Dimethyl-1-(phenylmethyl)-1H-pyrrol-3-yl]-2-oxoethoxy]phenyl]-2-furancarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N2O4
CAS Number:
Molecular Weight:
428.48
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

SA-15-P is a LAG-3 (CD223) inhibitor that blocks LAG-3 interactions with both major histocompatibility complex class II and fibrinogen-like protein 1 by cell-free (MHCII IC50 = 4.21 μM & FGL1 IC50 = 6.52 μM) and cell-based binding assays (IC50 ~15 μM against 150 ng/mL or 1.5 nM FGL1-hFc binding to stable LAG-3-expressing CHO-K1 cell line).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of First-in-Class Small Molecule Inhibitors of Lymphocyte Activation Gene 3 (LAG-3)
ACS Medicinal Chemistry Letters, 14(5), 629-635 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service