Skip to Content
Merck
All Photos(2)

Key Documents

SML3614

Sigma-Aldrich

Vatiquinone

≥95% (HPLC)

Synonym(s):

α-tocotrienol quinone, 2-[(3R,6E,10E)-3-Hydroxy-3,7,11,15-tetramethyl-6,10,14-hexadecatrien-1-yl]-3,5,6-trimethyl-2,5-cyclohexadiene-1,4-dione, ATQ3, EPI 743, EPI-743

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€183.00
25 MG
€772.00

€183.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€183.00
25 MG
€772.00

About This Item

Empirical Formula (Hill Notation):
C29H44O3
CAS Number:
Molecular Weight:
440.66
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

€183.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

oil

color

, Colorless to light yellow

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally available, highly potent analog of coenzyme Q10 that modulates the activity of oxidoreductases and improves cell redox status
Vatiquinone (EPI-743), a metabolite of a-tocotrienol, is an orally available and highly potent analog of coenzyme Q10 that modulates the activity of oxidoreductases including AD(P)H:quinone oxidoreductase 1, resulting in increased cellular GSH concentration and improvement in redox status. Vatiquinone is an agent designated to treat mitochondrial diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid identification of a novel complex I MT-ND3 m.10134C>A mutation in a Leigh syndrome patient
PLoS ONE, 9(8), e10487-e10487 (2014)
S Küpeli et al.
The Journal of urology, 141(2), 378-378 (1989-02-01)
We describe a patient with a radiopaque calculus in the vas deferens that was inappropriately diagnosed as a ureteral calculus.
William D Shrader et al.
Bioorganic & medicinal chemistry letters, 21(12), 3693-3698 (2011-05-24)
We report that α-tocotrienol quinone (ATQ3) is a metabolite of α-tocotrienol, and that ATQ3 is a potent cellular protectant against oxidative stress and aging. ATQ3 is orally bioavailable, crosses the blood-brain barrier, and has demonstrated clinical response in inherited mitochondrial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service