Skip to Content
Merck
All Photos(1)

Key Documents

SML2596

Sigma-Aldrich

Sphynolactone-7

≥98% (HPLC)

Synonym(s):

4-Methyl-5-oxo-2,5-dihydrofuran-2-yl 4-(4-butoxyphenylsulfonyl)piperazine-1-carboxylate, SPL-7, SPL7

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€118.00
25 MG
€363.00

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€118.00
25 MG
€363.00

About This Item

Empirical Formula (Hill Notation):
C20H26N2O7S
CAS Number:
Molecular Weight:
438.49
UNSPSC Code:
12352200
NACRES:
NA.77

€118.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCCOC1=CC=C(S(N2CCN(C(OC3C=C(C)C(O3)=O)=O)CC2)(=O)=O)C=C1

InChI key

GGAPQPVAXJSNFU-UHFFFAOYSA-N

Biochem/physiol Actions

Highly potent & selective strigolactone receptor ShHTL7 agonist. Femto-/Pico-molar germination stimulant of the parasitic plant Striga hermonthica/Orobanche minor.
Sphynolactone-7 (SPL-7) is an active site-targeting, highly potent and selective strigolactone receptor ShHTL7 agonist (YLG competitive binding IC50 = 310 nM/ShHTL7, 1.2 μM/ShHTL8, >7.0 μM/ShHTL2-6, ShHTL9-11, AtD14) that acts as a seeds germination stimulant with femtomolar- and picomolar-level potency, respectively, toward the parasitic plants Striga hermonthica ecotypes and Orobanche minor, while exhibiting no Arabidopsis-stimulatory efficacy.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Uraguchi et al.
Science (New York, N.Y.), 362(6420), 1301-1305 (2018-12-14)
The parasitic plant Striga hermonthica has been causing devastating damage to the crop production in Africa. Because Striga requires host-generated strigolactones to germinate, the identification of selective and potent strigolactone agonists could help control these noxious weeds. We developed a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service