Skip to Content
Merck
All Photos(1)

Key Documents

SML2419

Sigma-Aldrich

GSK690

≥98% (HPLC)

Synonym(s):

(R)-4-(5-(Pyrrolidin-3-ylmethoxy)-2-(p-tolyl)pyridin-3-yl) benzonitrile), (R)-4-(5-(Pyrrolidin-3-ylmethoxy)-2-p-tolylpyridin-3-yl)benzonitrile, 4-(2-(4-Methylphenyl)-5-((3R)-3-pyrrolidinylmethoxy)-3-pyridinyl)benzonitrile, GSK 354, GSK 690, GSK-354, GSK-690, GSK354

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€65.60
25 MG
€265.00

€65.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€65.60
25 MG
€265.00

About This Item

Empirical Formula (Hill Notation):
C24H23N3O
CAS Number:
Molecular Weight:
369.46
UNSPSC Code:
12352200

€65.60


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=CC=C(C2=CC(OC[C@H]3CNCC3)=CN=C2C4=CC=C(C)C=C4)C=C1

InChI

1S/C24H23N3O/c1-17-2-6-21(7-3-17)24-23(20-8-4-18(13-25)5-9-20)12-22(15-27-24)28-16-19-10-11-26-14-19/h2-9,12,15,19,26H,10-11,14,16H2,1H3/t19-/m1/s1

InChI key

IQVDLEXWAPYWDT-LJQANCHMSA-N

Biochem/physiol Actions

GSK690 (GSK354) is a cell-permeable pyrrolidinylmethoxypyridine-based compound that inhibits lysine-specific demethylase 1 (LSD1; KDM1A) in a potent and reversible manner (IC50 = 90 nM) without affecting MAO-A activity (IC50 >200 μM). GSK690 is shown to induce the expressions of CD86 (EC50 = 1.4 μM in THP1 cells) and LY96 (EC50 = 423 nM in MV4-11 cells), and prevent adipogenesis in murine preadipocyte cultures.
Potent and reversible lysine-specific demethylase LSD1 (KDM1A) inhibitor that induces CD86 and LY96 expression and prevents adipogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel P Mould et al.
Journal of medicinal chemistry, 60(19), 7984-7999 (2017-09-12)
Inhibition of lysine specific demethylase 1 (LSD1) has been shown to induce the differentiation of leukemia stem cells in acute myeloid leukemia (AML). Irreversible inhibitors developed from the nonspecific inhibitor tranylcypromine have entered clinical trials; however, the development of effective
Alba Maiques-Diaz et al.
Cell reports, 22(13), 3641-3659 (2018-03-29)
Pharmacologic inhibition of LSD1 promotes blast cell differentiation in acute myeloid leukemia (AML) with MLL translocations. The assumption has been that differentiation is induced through blockade of LSD1's histone demethylase activity. However, we observed that rapid, extensive, drug-induced changes in transcription occurred
Tinka Haydn et al.
Cell death & disease, 8(6), e2879-e2879 (2017-06-16)
The lysine-specific demethylase 1 (LSD1) is overexpressed in several cancers including rhabdomyosarcoma (RMS). However, little is yet known about whether or not LSD1 may serve as therapeutic target in RMS. We therefore investigated the potential of LSD1 inhibitors alone or
Development and evaluation of selective, reversible LSD1 inhibitors derived from fragments
Hitchin JR, Blagg J, Burke R .
Medicinal Chemistry, 1513-1522 (2013)
Delphine Duteil et al.
Nature communications, 5, 4093-4093 (2014-06-11)
Exposure to environmental cues such as cold or nutritional imbalance requires white adipose tissue (WAT) to adapt its metabolism to ensure survival. Metabolic plasticity is prominently exemplified by the enhancement of mitochondrial biogenesis in WAT in response to cold exposure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service