SML1838
BNS
≥98% (HPLC)
Synonym(s):
2,3-Dihydro-3-hydroxy-2-[[2-[(2-naphthalenylmethyl)sulfonyl]acetyl]imino]-4-thiazoleacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 20 mg/mL, clear
storage temp.
2-8°C
SMILES string
ON1C(CC(O)=O)=CSC1=NC(CS(CC2=CC=C(C=CC=C3)C3=C2)(=O)=O)=O
Biochem/physiol Actions
BNS is a cell penetrant, potent and selective inhibitor of prolyl-hydroxylase 2 (PHD2). BNS regulates ~25% of hypoxia-regulated genes in MCF7 cells.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bioorganic & medicinal chemistry letters, 18(14), 3925-3928 (2008-06-27)
Inhibition of the PHD2 enzyme has been associated with increased red blood cell levels. From a screening hit, a series of novel hydroxyl-thiazoles were developed as potent PHD2 inhibitors.
The Journal of biological chemistry, 291(39), 20661-20673 (2016-08-10)
The hypoxia-inducible factor (HIF) system orchestrates cellular responses to hypoxia in animals. HIF is an α/β-heterodimeric transcription factor that regulates the expression of hundreds of genes in a tissue context-dependent manner. The major hypoxia-sensing component of the HIF system involves
Journal of medicinal chemistry, 56(2), 547-555 (2012-12-14)
The human 2-oxoglutarate (2OG) dependent oxygenases belong to a family of structurally related enzymes that play important roles in many biological processes. We report that competition-based NMR methods, using 2OG as a reporter ligand, can be used for quantitative and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service