Skip to Content
Merck
All Photos(1)

Key Documents

SML1701

Sigma-Aldrich

BX430

≥98% (HPLC)

Synonym(s):

N-[2,6-Dibromo-4-(1-methylethyl)phenyl]-N′-(3-pyridinyl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€81.60
25 MG
€334.00

€81.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€81.60
25 MG
€334.00

About This Item

Empirical Formula (Hill Notation):
C15H15Br2N3O
CAS Number:
Molecular Weight:
413.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€81.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=CC(C(C)C)=CC(Br)=C1NC(NC2=CC=CN=C2)=O

InChI

1S/C15H15Br2N3O/c1-9(2)10-6-12(16)14(13(17)7-10)20-15(21)19-11-4-3-5-18-8-11/h3-9H,1-2H3,(H2,19,20,21)

InChI key

JFNKIJKRXKPQCC-UHFFFAOYSA-N

Biochem/physiol Actions

BX430 is a selective noncompetitive allosteric antagonist of human P2X4 receptor channels.
BX430 is a selective noncompetitive allosteric antagonist of human P2X4 receptor channels. P2X4 receptors are highly expressed in the CNS, and have been studied as a therapeutic target for neuropathic pain and inflammation, and treatment of traumatic brain injury, cerebral ischemia, and spinal cord injury. BX430 is highly selective for human P2X4, with minimal activity towards other P2X subtypes, including P2X1–P2X3, P2X5, and P2X7. BX430 is also an antagonist of zebrafish P2X4 but has no effect on rat and mouse P2X4 receptors. BX430 has an IC50 value of 540 nM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service